missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phosphate-buffered saline (PBS, 5X), with EDTA

Product Code. p-7082247
Click to view available options
Quantity:
1 L
2 L
Unit Size:
1 L
2 L
This item is not returnable. View return policy

Product Code. 15433589

Brand: Thermo Scientific Chemicals J60893.K2

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Helps to maintain a constant pH

PBS with EDTA is also used to disengage attached and clumped cells. PBS-EDTA is used as a buffer in procedures generating dendritic cells from monocytes. PBS+EDTA to get better cell sorting. PBS/EDTA may also be used directly without addition of trypsine for very gentle cell dissociation. PBS can be used to take a reference spectrum when measuring the protein adsorption in ellipsometry. PBS can be used as a diluent in methods to dry biomolecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Concentration 5 X
Physical Form Liquid
Quantity 1 L

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.