missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Phosphate Buffered Saline Tablet
Each tablet prepares 100mL of 1X PBS solution. Fisher Bioreagents™ Phosphate Buffered Saline tablets (PBS) are a convenient option for making 1X phosphate buffered saline.
Brand: Fisher BioReagents™ 12821680
This item is not returnable.
View return policy
Description
Tablet formulation:- Sodium Chloride: 137mM
- Phosphate Buffer: 10mM
- Potassium Chloride: 2. 7mM
Specifications
1 X | |
Phosphate Buffered Saline (PBS) | |
1 g | |
7.3 to 7.5 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- PBS Tablets
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Potassium Chloride,Sodium Chloride, Phosphate Buffer
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
Days
0Hours
10Minutes
48Seconds
48
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisher BioReagents™ Phosphate Buffered Saline Tablet > Phosphate Buffered Saline Tablet
Spot an opportunity for improvement?Share a Content Correction