missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Phospho-Akt (S473) Pan Specific DuoSet IC ELISA
Phospho-Akt (S473) Pan Specific DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand: R&D Systems™ DYC887B-2
This item is not returnable.
View return policy
Description
Complete ELISpot kits are highly sensitive, microplate-based assays for the detection of cytokine secreting cells. Kits are available for detection and enumeration of a single analyte or two analytes simultaneously.
The Phospho-Akt (S473) Pan Specific DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human, mouse, rat AKT.Specifications
ELISA Kit | |
Human, Mouse, Rat | |
156.00 to 10000 pg/mL | |
2 96-well Plates | |
RAC-ALPHA, AKT, AKT serine/threonine kinase 1, PKB, PKB alpha, PKB beta, PKB gamma, PKB-ALPHA, PRKBA, RAC, RAC-alpha, RAC-beta, RAC-gamma |
1 Kit (for 2 Plates) | |
Biotin | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules | |
ELISA | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |