missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-AKT (Ser473) Mouse anti-Human, Mouse, Clone: 1C10B8, Proteintech
Mouse Monoclonal Antibody
£120.00 - £271.00
Specifications
Antigen | Phospho-AKT (Ser473) |
---|---|
Clone | 1C10B8 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16878325
|
Proteintech
66444-1-IG-20UL |
20 μL |
£120.00
20µL |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16868325
|
Proteintech
66444-1-IG-100UL |
100 μL |
£271.00
100µL |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody detects all the members of AKT with phospho-modification at Ser473.
The Akt family of serine/threonine protein kinases (also called protein kinase B/PKB) plays a key role in insulin signaling, cellular survival, and transformation. Akt1 is activated by phosphorylation on residues threonine 308 and Serine 473. Thr308, in the catalytic domain, is phosphorylated by the protein kinase PDK1, and Ser473, in the COOH-terminal regulatory domain, is phosphorylated by putative serine 473 kinase. To date, a couple of proteins have been shown to physically interact with PKB.Specifications
Phospho-AKT (Ser473) | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
AKT, AKT1, AKT1-phospho-S473, pakt, p-AKT, Phospho akt, PKB, PKB ALPHA, PRKBA, Protein kinase B, Proto oncogene c Akt, RAC, RAC ALPHA, RAC PK alpha | |
AKT1 | |
IgG1 | |
Protein G | |
Antibody |
1C10B8 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
P31749, P31750 | |
11651, 207 | |
Peptide | |
Primary | |
-20°C | |
AKT1 |