missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-ATR (Ser428) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5121295
Additional Details : Weight : 0.01000kg
Description
Positive test controls include: NIH/3T3. The target is usually found in the following locations: Chromosome, Nucleus, PML body. Immunogen sequence: DGISPK
The protein encoded by this gene is a serine/threonine kinase and DNA damage sensor, activating cell cycle checkpoint signaling upon DNA stress. The encoded protein can phosphorylate and activate several proteins involved in the inhibition of DNA replication and mitosis, and can promote DNA repair, recombination, and apoptosis. This protein is also important for fragile site stability and centrosome duplication. Defects in this gene are a cause of Seckel syndrome 1.Specifications
Phospho-ATR (Ser428) | |
Polyclonal | |
Unconjugated | |
ATR | |
2310008J16Rik; 2810405N18Rik; ataxia telangiectasia and Rad3 related; ataxia telangiectasia and Rad3-related protein; ATR; ATR serine/threonine kinase; EC 2.7.11.1; FCTCS; FRAP-related protein 1; FRAP-related protein-1; FRP1; I79_000761; kinase ATR; MEC1; MEC1, mitosis entry checkpoint 1, homolog; protein kinase ATR; SCKL; SCKL1; Serine/threonine-protein kinase ATR; TEM8 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
245000, 545, 685055 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q13535, Q9JKK8 | |
ATR | |
A synthetic phosphorylated peptide around S428 of human ATR (NP_001175.2). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |