missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-eIF4G (Ser1108) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA517824
Code : LE
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with nonphosphorylated eIF4GI or p97.
The protein encoded by this gene is a component of the protein complex EIF4F, which is involved in the recognition of the mRNA cap, ATP-dependent unwinding of 5'-terminal secondary structure, and recruitment of mRNA to the ribosome. Alternative splicing results in five transcript variants encoding four distinct isoforms.Specifications
Phospho-eIF4G (Ser1108) | |
Polyclonal | |
Unconjugated | |
EIF4G1 | |
DKFZp686A1451; E030015G23Rik; EIF4F; EIF4G; eIF-4G 1; EIF4G1; eIF-4G1; EIF4-gamma; eIF-4-gamma 1; EIF4GI; eIF4GI protein; eucaryotic translation initiation factor 4G; Eukaryotic translation initiation factor 4 gamma 1; eukaryotic translation initiation factor 4 gamma, 1; eukaryotic translation initiation factor 4, gamma 1; OTTHUMP00000210577; OTTHUMP00000210586; OTTHUMP00000210587; OTTHUMP00000210588; OTTHUMP00000210595; OTTHUMP00000210602; OTTHUMP00000210603; P220; PARK18 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100770567, 1981, 208643, 287986, 444858 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
142 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
Q04637, Q6NZJ6 | |
EIF4G1 | |
Synthetic phosphopeptide corresponding to residues surrounding pSer1108 of human eIF4GI | |
100 μL | |
Primary | |
Bovine, Hamster, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |