Learn More
Phospho-eNOS (Ser1177) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA535879
Code : NEW
Additional Details : Weight : 0.00910kg
Description
Recommended positive controls: Raji, NCI-H929, THP-1, THP-1 (100 nM PMA treatment for 6, 24, 48 hr). Predicted reactivity: Mouse (100%), Rat (100%), Dog (100%), Pig (100%), Rabbit (100%), Sheep (100%), Bovine (100%), Guinea pig (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Phospho-eNOS (Ser1177) Polyclonal antibody specifically detects Phospho-eNOS (Ser1177) in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Phospho-eNOS (Ser1177) | |
Polyclonal | |
Unconjugated | |
NOS3 | |
2310065A03Rik; cNOS; Constitutive NOS; Ecnos; EC-NOS; endothelial nitric oxide synthase; endothelial nitric oxide synthase 3; endothelial nitric oxide synthase NOS3; endothelial NOS; eNos; e-NOS; nitric oxide synthase; nitric oxide synthase 3; nitric oxide synthase 3 (endothelial cell); nitric oxide synthase 3 transcript variant eNOS-delta20; nitric oxide synthase 3 transcript variant eNOS-delta20-21; nitric oxide synthase 3 transcript variant eNOS-delta21; nitric oxide synthase 3, endothelial cell; nitric oxide synthase, endothelial; NOS; NOS type III; Nos3; Nos-3; NOSIII; OTTHUMP00000213183 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18127, 24600, 4846 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.94 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
P29474, P70313, Q62600 | |
NOS3 | |
Synthetic phospho-peptide corresponding to amino acids residues surrounding the phospho Ser1177 of eNOS | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |