missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-ENT1 (Ser254) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA513044
Code : LE
Additional Details : Weight : 0.01000kg
Description
This gene is a member of the equilibrative nucleoside transporter family. The gene encodes a transmembrane glycoprotein that localizes to the plasma and mitochondrial membranes and mediates the cellular uptake of nucleosides from the surrounding medium. The protein is categorized as an equilibrative transporter that is sensitive to inhibition by nitrobenzylthioinosine. Nucleoside transporters are required for nucleotide synthesis in cells that lack de novo nucleoside synthesis pathways, and are also necessary for the uptake of cytotoxic nucleosides used for cancer and viral chemotherapies. Multiple alternatively spliced variants, encoding the same protein, have been found for this gene.Specifications
Phospho-ENT1 (Ser254) | |
Polyclonal | |
Unconjugated | |
SLC29A1 | |
1200014D21Rik; AA407560; ENT1; Equilibrative NBMPR-sensitive nucleoside transporter; equilibrative nitrobenzylmercaptopurine riboside (NBMPR)-sensitive nucleoside transporter; equilibrative nitrobenzylmercaptopurine riboside-sensitive nucleoside transporter; Equilibrative nucleoside transporter 1; equilibrative nucleoside transporter 1 variant delta 11; HENT1; NBMPR-sensitive equilibrative nucleoside transporter; nucleoside transporter, es-type; rENT1; SLC29A1; solute carrier family 29 (equilibrative nucleoside transporter), member 1; solute carrier family 29 (nucleoside transporters), member 1; solute carrier family 29 member 1; solute carrier family 29 member 1 (Augustine blood group); solute carrier family 29, member 1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
2030 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Dot Blot | |
0.25 mg/mL | |
PBS with 0.09% sodium azide | |
Q99808 | |
SLC29A1 | |
KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S254 of human ENT1(Slc29a1) | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |