missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-FANCD2 (Ser330) Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£112.27 - £256.47
Specifications
Antigen | Phospho-FANCD2 (Ser330) |
---|---|
Concentration | 0.37 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16899074
|
Proteintech
19543-1-AP-20UL |
20 μL |
£112.27
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16889074
|
Proteintech
19543-1-AP-100UL |
100 μL |
£256.47
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The antibody is specific to phospho-Ser(330) of FANCD2.
Fanconi anemia (FANC) is a human autosomal-recessive cancer susceptibility disorder characterized by congenital defects, progressive bone marrow failure, and cellular hypersensitivity to mitomycin C (MMC). The FANC subunit D2 protein is vital for cellular resistance to DNA cross-linking and the arrest of DNA synthesis after ionizing radiation. DNA damage activates the monoubiquitination of FANCD2, targeting nuclear foci containing the BRCA1 protein.Specifications
Phospho-FANCD2 (Ser330) | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q9BXW9 | |
2177 | |
Peptide | |
Primary | |
-20°C | |
FANCD2 |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
FA D2, FA4, FACD, FAD, FAD2, FANCD, FANCD2, Protein FACD2 | |
FANCD2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |