missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-NBS1 (Ser343) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578070
This item is not returnable.
View return policy
Description
Positive Control: HeLa, HeLa (100 J/M2 UVC treatment and recovery for 4 hr) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The p95 gene (identical to NBS1 and nibrin) is a member of the hMre11/hRad50 double-strand break complex. This protein complex has been implicated in Nijmegen breakage syndrome, an autosomal recessive disorder marked by increased cancer incidence, cell cycle checkpoint deficits, and ionizing radiation sensitivity, thus revealing a direct molecular link between double-strand break repair and cell cycle checkpoint functions.
Specifications
| Phospho-NBS1 (Ser343) | |
| Polyclonal | |
| Unconjugated | |
| NBN | |
| AT V1; AT V2; ATV; AT-V1; AT-V2; Cell cycle regulatory protein p95; FLJ10155; I79_012607; MGC87362; Nbn; NBS; NBS1; nibrin; Nijmegen breakage syndrome 1 (nibrin); Nijmegen breakage syndrome protein 1; Nijmegen breakage syndrome protein 1 homolog; OTTHUMP00000204088; P95; p95 protein of the MRE11/RAD50 complex; p95-NBS1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4683 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.97 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
| O60934 | |
| NBN | |
| Carrier-protein conjugated synthetic peptide surrounding phospho Ser343 of human NBS1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction