missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-PI3K p85/p55 (Tyr467, Tyr199) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538903
This item is not returnable.
View return policy
Description
Phosphatidylinositol 3-kinase phosphorylates the inositol ring of phosphatidylinositol at the 3-prime position. The enzyme comprises a 110kDa catalytic subunit and a regulatory subunit of either 85, 55, or 50kDa. It binds to activated (phosphorylated) protein-Tyr kinases, through its SH2 domain, and acts as an adapter, mediating the association of the p110 catalytic unit to the plasma membrane. Phosphatidylinositol 3-kinase plays an important role in the metabolic actions of insulin, and a mutation in this gene has been associated with insulin resistance. During insulin stimulation, it also binds to IRS-1.Specifications
Phospho-PI3K p85/p55 (Tyr467, Tyr199) | |
Polyclonal | |
Unconjugated | |
PIK3R3 | |
Phosphatidylinositol 3-kinase regulatory subunit alpha, PI3-kinase regulatory subunit alpha, PI3K regulatory subunit alpha, PtdIns-3-kinase regulatory subunit alpha, Phosphatidylinositol 3-kinase 85 kDa regulatory subunit alpha, PI3-kinase subunit p85-alpha, PtdIns-3-kinase regulatory subunit p85-al, RP4-533D7.2, p55, p55-GAMMA, 100% homology to SWISS-PROT Q92569, PI3-kinase regulatory subunit gamma, PI3-kinase subunit p55-gamma, PI3K regulatory subunit gamma, p55PIK, phosphatidylinositol 3-kinase 55 kDa regulatory subunit gamma, phosphatidylinositol 3-kinase regulatory subunit gamma, phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 3 (p55, gamma), phosphoinositide-3-kinase, regulatory subunit 3 (p55, gamma), phosphoinositide-3-kinase, regulatory subunit, polypeptide 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18708, 18710, 25513, 5295, 60664, 8503 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P27986, Q92569, P26450, Q64143, Q63787, Q63789 | |
PIK3R1, PIK3R3 | |
A synthetic phosphopeptide derived from human PI3-kinase p85-subunit alpha/gamma around the phosphorylation site of Tyr467/199 (R-L-YP-E-E) | |
100 μg | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction