missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Smtnl1 (Ser301) Rabbit anti-Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£112.27 - £256.47
Specifications
Antigen | Phospho-Smtnl1 (Ser301) |
---|---|
Concentration | 0.22 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16899094
|
Proteintech
19560-1-AP-20UL |
20 μL |
£112.27
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16889094
|
Proteintech
19560-1-AP-100UL |
100 μL |
£256.47
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The antibody recognize Ser(301) phospho of Smtn1.
Plays a role in the regulation of contractile properties of both striated and smooth muscles. When unphosphorylated, may inhibit myosin dephosphorylation. Phosphorylation at Ser-299 reduces this inhibitory activity.Specifications
Phospho-Smtnl1 (Ser301) | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse | |
Q99LM3 | |
68678 | |
Peptide | |
Primary | |
-20°C | |
Smtnl1 |
0.22 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
1110030K22Rik, CHASM, smoothelin like 1, Smtnl1 | |
Smtnl1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |