missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoenolpyruvic acid monopotassium salt, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Phosphoenolpyruvic acid monopotassium salt is used in glycolysis and gluconeogeneis. It is used for regeneration of ATP from ADP in enzyme-catalyzed syntheses. It is metabolized by an enzyme namely pyruvate kinase to yield pyruvate. It acts as protection against cell injury induced by oxidative stress and against dysfunction in carbohydrate metabolism.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phosphoenolpyruvic Acid Monopotassium Salt is used in glycolysis and gluconeogeneis. It is used for regeneration of ATP from ADP in enzyme-catalyzed syntheses. It is metabolized by an enzyme namely pyruvate kinase to yield pyruvate. It acts as protection against cell injury induced by oxidative stress and against dysfunction in carbohydrate metabolism.
Solubility
Soluble in water. Slightly soluble in methanol.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Phosphoenolpyruvic Acid Monopotassium Salt is used in glycolysis and gluconeogeneis. It is used for regeneration of ATP from ADP in enzyme-catalyzed syntheses. It is metabolized by an enzyme namely pyruvate kinase to yield pyruvate. It acts as protection against cell injury induced by oxidative stress and against dysfunction in carbohydrate metabolism.
Solubility
Soluble in water. Slightly soluble in methanol.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | ∽170°C (decomposition) |
Assay Percent Range | 0.99 |
Quantity | 5 g |
Beilstein | 4603446 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. Slightly soluble in methanol. |
Formula Weight | 206.14 |
Percent Purity | 99% |
Chemical Name or Material | Phosphoenol pyruvate monopotassium salt |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'