missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Phosphogluconic Acid
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
Brand: Melford P20350-0.5
This item is not returnable.
View return policy
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
- CAS Number: 53411-70-4
- Molecular Formula: C6H10O10PNa3 2H2O
- Molecular Weight: 378.11
Specifications
6-Phosphogluconic Acid Trisodium Salt Dihydrate | |
57775-17-4 | |
C6H10Na3O10P-2H2O | |
500 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction