missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphomolybdic acid, ammonium salt hydrate, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Yellow |
Packaging | Plastic bottle |
Quantity | 50 g |
Linear Formula | (NH4)3PMo12O40·xH2O |
Physical Form | Powder |
Solubility Information | Solubility in water: insoluble. |
Formula Weight | 1876.35 |
Chemical Name or Material | Phosphomolybdic acid, ammonium salt hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'