missing translation for 'onlineSavingsMsg'
Learn More

Phosphomolybdic Acid Hydrate, Honeywell Fluka™

Product Code. 15612160
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51429-74-4
Molecular Formula H3Mo12O40P
Molecular Weight (g/mol) 1825.40
MDL Number MFCD00149913
InChI Key FEFSFHGZSNHJEL-UHFFFAOYSA-N
SMILES [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].OP(O)(O)=O

Specifications

Density 1.62g/mL at 25°C
Quantity 100 g
Linear Formula H3[P(Mo3O10)4] · xH2O
UN Number 3084
IUPAC Name phosphoric acid dodecamolybdenum hexatriacontaoxidandiide
Formula Weight 1825.25g/mol (anhydrous basis)
Grade ACS Reagent
Chemical Name or Material Phosphomolybdic Acid Hydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'