missing translation for 'onlineSavingsMsg'
Learn More
Phosphomolybdotungstic Reagent, Reagecon™ Product Code.: 11434719
compliance-icons

Phosphomolybdotungstic Reagent, Reagecon™
BIOPHARMA

Product Code. 11434719 Shop All Reagecon Products
100 mL
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy

Product Code. 11434719

Brand: Reagecon 1065000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Quantity 100 mL

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number16524L1 Certificate TypeCertificate of Analysis Date16/01/2025 Catalog Number1065000
Lot Number16524G1 Certificate TypeCertificate of Analysis Date24/09/2024 Catalog Number1065000
Lot Number16524F1 Certificate TypeCertificate of Analysis Date07/08/2024 Catalog Number1065000
Lot Number16524E1 Certificate TypeCertificate of Analysis Date21/06/2024 Catalog Number1065000
Lot Number16523M1 Certificate TypeCertificate of Analysis Date15/03/2024 Catalog Number1065000
compliance-icons
Product Identifier
  • phosphomolybdotungstic reagent
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phosphomolybdotungstic Reagent, Reagecon™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.