missing translation for 'onlineSavingsMsg'
Learn More
Phosphonitrilic chloride trimer, 98%, Thermo Scientific Chemicals Product Code.: 11419306

Phosphonitrilic chloride trimer, 98%, Thermo Scientific Chemicals

Product Code. 11419306
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11419306

Brand: Thermo Scientific Alfa Aesar A18986.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 940-71-6
Molecular Formula Cl6N3P3
Molecular Weight (g/mol) 347.642
MDL Number MFCD00006474
InChI Key UBIJTWDKTYCPMQ-UHFFFAOYSA-N
Synonym phosphonitrilic chloride trimer, hexachlorocyclotriphosphazene, hexachlorophosphazene, triphosphonitrilic chloride, triphosphonitrile chloride, hexachlorotriphosphonitrile, cyclophosphazene dichloride trimer, hexachlorocyclophosphazatriene, hexachlorocyclotriphosphazatriene, phosphononitrilic chloride trimerShow MoreShow Less
PubChem CID 220225
IUPAC Name 2,2,4,4,6,6-hexachloro-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene
SMILES N1=P(N=P(N=P1(Cl)Cl)(Cl)Cl)(Cl)Cl

Specifications

Boiling Point 127°C (13mmHg)
Melting Point 111°C to 115°C
Quantity 100 g
UN Number UN3260
Formula Weight 347.66
Percent Purity 98%
Sensitivity Moisture sensitive
Density 1.98
Chemical Name or Material Phosphonitrilic chloride trimer

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10248685 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog NumberA18986.22,A18986.14,A18986.06
Lot Number10245331 Certificate TypeCertificate of Analysis Date13/01/2023 Catalog NumberA18986.22,A18986.14,A18986.06
Lot Number10240008 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA18986.22,A18986.14,A18986.06
Lot Number10237653 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA18986.22,A18986.14,A18986.06
Lot Number10148109 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18986.22,A18986.14,A18986.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phosphonitrilic chloride trimer, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.