missing translation for 'onlineSavingsMsg'
Learn More
Phosphoric acid, 85% w/w aq. soln., ACS, Thermo Scientific™ Product Code.: 11337578

Phosphoric acid, 85% w/w aq. soln., ACS, Thermo Scientific™

Product Code. 11337578
100 g
Click to view available options
Quantity:
100 g
1 kg
4 x 1 kg
This item is not returnable. View return policy

Product Code. 11337578

Brand: Thermo Scientific Alfa Aesar 033266.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Phosphoric acid is used mainly in fertilizers. It is used as an external standard for phosphorus-31 Nuclear magnetic resonance (NMR); as an electrolyte in phosphoric acid fuel cells and as a catalyst in the hydration of alkenes to produce alcohols, predominantly ethanol. It finds application as a buffer agent in biology and chemistry; as a solution for anodizing and as a cleaner by construction trades to remove mineral deposits, cementitious smears and hard water stains. It is also used as a pH adjuster in cosmetics and skin-care products and a dispersing agent in detergents and in leather treatment. It plays an important role in the preparation of albumin derivatives. It is an important starting material used in the preparation of disodium hydrogen phosphate, trisodium phosphate, calcium dihydrogen phosphate and monohydrogen phosphate. It serves as an acidic fruitlike flavoring in food products. It is widely used to remove rust in steel tools or surfaces by converting the rusted ion into a water soluble phosphate compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
MDL Number MFCD00011340
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforicoShow MoreShow Less
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Melting Point 19°C to 21°C
Color Colorless
Physical Form Viscous Liquid
Quantity 100 g
Assay Percent Range 85% w/w aq. soln.
Solubility Information Miscible with water,ethanol and mixture of ether: alcohol (3:1).
Formula Weight 98
Grade ACS
Refractive Index 1.431
Appearance Colorless viscous liquid
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Density 1.70g/cm3 at 20°C
Chemical Name or Material Phosphoric acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN14L025 Certificate TypeCertificate of Analysis Date17/02/2025 Catalog Number033266.D6,033266.22,033266.A1
Lot NumberM03L017 Certificate TypeCertificate of Analysis Date21/01/2025 Catalog Number033266.D6,033266.22,033266.A1
Lot NumberX15K001 Certificate TypeCertificate of Analysis Date25/10/2024 Catalog Number033266.D6,033266.22,033266.A1
Lot NumberQ17K039 Certificate TypeCertificate of Analysis Date23/04/2024 Catalog Number033266.D6,033266.22,033266.A1
Lot NumberP06K010 Certificate TypeCertificate of Analysis Date13/03/2024 Catalog Number033266.D6,033266.22,033266.A1
compliance-icons compliance-icons
Product Identifier
  • Phosphoric acid, 85% w/w aqueous solution, ACS
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Phosphoric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phosphoric acid, 85% w/w aq. soln., ACS, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.