missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric acid, for analysis, 85 wt% solution in water, Thermo Scientific Chemicals

£38.69 - £284.42

Chemical Identifiers

CAS 7664-38-2, 7732-18-5
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
MDL Number MFCD00011340
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10685082
View Documents
Thermo Scientific Acros
201145000
500 mL Glass bottle
£38.69
500mL
Please to purchase this item. Need a web account? Register with us today!
10318150
View Documents
Thermo Scientific Acros
201140010
1 L Glass bottle
£58.79
1 L
Please to purchase this item. Need a web account? Register with us today!
10216690
View Documents
Thermo Scientific Acros
201140025
2.5 L Glass bottle
£93.97
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10458283
View Documents
Thermo Scientific Acros
201140100
10 L Plastic drum
£284.42
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7664-38-2, 7732-18-5
97.994
NBIIXXVUZAFLBC-UHFFFAOYSA-N
1004
phosphoric acid
H3O4P
MFCD00011340
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
CHEBI:26078
OP(=O)(O)O

Specifications

7664-38-2, 7732-18-5
158.0°C
1.68
H3PO4
MFCD00011340
15, 7456
Solubility in water: miscible.
OP(=O)(O)O
97.994
CHEBI:26078
≥85%
Phosphoric acid, 85 wt% Solution in Water
21.0°C
1.6800g/mL
H3O4P
Oily Liquid
01,860; 04,387; 07,255; 10,317; 15,266; 16,177
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
NBIIXXVUZAFLBC-UHFFFAOYSA-N
phosphoric acid
1004
98
Analytical
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

Product Identifier
  • Phosphoric acid, 85+% solution in water

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4

Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Orthophosphoric acid

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only