missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric acid, pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | White |
Melting Point | 42.0°C |
Density | 1.68 |
Packaging | Glass bottle |
Quantity | 500 g |
Linear Formula | H3PO4 |
Physical Form | Crystals or Crystalline Powder |
Merck Index | 15, 7456 |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble ethanol |
Formula Weight | 98 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0468024 | Certificate of Analysis | 31/01/2025 | 389021000,389025000,389020025 |
A0466795 | Certificate of Analysis | 30/01/2025 | 389021000,389025000,389020025 |
A0464785 | Certificate of Analysis | 23/07/2024 | 389021000,389025000,389020025 |
A0463838 | Certificate of Analysis | 14/06/2024 | 389021000,389025000,389020025 |
A0427691 | Certificate of Analysis | 21/09/2023 | 389021000,389025000,389020025 |
Safety and Handling

Product Identifier
- Phosphoric acid
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Phosphoric acid, pure, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction