missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric acid, Puriss., meets analytical spec. of BP, FCC, NF, Ph. Eur., 85.0-88.0%, Honeywell Fluka™
BIOPHARMA

Product Code. 15670710 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
4 x 2.5 L
45 kg
4 x 5 L
6 x 1 L
Unit Size:
1 L
10 L
2.5 L
45kg
5 L
Pack of 4
Pack of 6
This item is not returnable. View return policy

Product Code. 15670710

Brand: Honeywell-Fluka 041074X2.5L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, FCC, NF, Ph. Eur., 85.0-88.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
MDL Number MFCD00011340
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Density 1.685g/mL at 25°C
Vapor Pressure 5mmHg (25°C), 2.2mmHg (20°C)
Quantity 4 x 2.5 L
Linear Formula H3PO4
UN Number UN1805
Formula Weight 98
Percent Purity 85.0 to 88.0%
Grade puriss.
Chemical Name or Material Phosphoric Acid, Meets analytical specification of Ph. Eur.BP, NF, FCC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.