missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥85%, Honeywell Fluka™
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
45 kg
6 x 1 L
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Melting Point | ∼40°C (literature) |
Boiling Point | 158°C |
CAS Min % | ≥70.0000%, ≥10.0000% |
CAS Max % | <90.0000%, <20.0000% |
Density | 1.685 g/mL at 25 °C(lit.) |
Packaging | Poly Bottle |
Quantity | 4 x 2.5 L |
Linear Formula | H3PO4 |
UN Number | UN1805 |
Beilstein | 1921286 |
Show More |
Safety and Handling
Product Identifier
- Phosphoric acid
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Orthophosphoric acid
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'