missing translation for 'onlineSavingsMsg'
Learn More
Phosphoric acid, Puriss. p.a., crystallized, ≥99.0% (T), Honeywell Fluka™ Product Code.: 15627160

Phosphoric acid, Puriss. p.a., crystallized, ≥99.0% (T), Honeywell Fluka™

Product Code. 15627160 Shop All Honeywell Research Chemicals Products
500 g, Glass Bottle
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 15627160

Brand: Honeywell-Fluka 79622500G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., crystallized,≥99.0% (T)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
MDL Number MFCD00011340
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Melting Point ∼40°C (literature)
Boiling Point 158°C (literature)
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Density 1.685 g/mL at 25 °C(lit.)
Packaging Glass Bottle
Vapor Pressure 2.2 mmHg (20°C), 5 mmHg (25°C)
Quantity 500 g
Linear Formula H3PO4
UN Number UN3453
Beilstein 1921286
Formula Weight 98
Percent Purity ≥99%
Grade puriss. p.a.
Chemical Name or Material Phosphoric acid, Puriss. p.a., crystallized, ≥99.0% (T)
Show More Show Less
compliance-icons
Product Identifier
  • Phosphoric acid
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Orthophosphoric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phosphoric acid, Puriss. p.a., crystallized, ≥99.0% (T), Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.