missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric Acid Solution, 33% (w/w), Biotech Reagent, J.T. Baker™
1135.25 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
19 L
Packaging:
Poly Bottle
Specifications
CAS | 7664-38-2 |
Packaging | Poly Bottle |
Specific Gravity | 1L = 1.20kg |
Quantity | 19 L |
Molecular Formula | H3O4P |
Synonym | orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico |
InChI Key | NBIIXXVUZAFLBC-UHFFFAOYSA-N |
SMILES | OP(=O)(O)O |
IUPAC Name | phosphoric acid |
Molecular Weight (g/mol) | 97.994 |
Show More |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'