missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric acid, TraceSELECT™, Honeywell Fluka™
TraceSELECT™, for trace analysis, ∼85%
£112.75 - £405.76
Chemical Identifiers
CAS | 7664-38-2 |
---|---|
Molecular Formula | H3O4P |
Molecular Weight (g/mol) | 97.994 |
MDL Number | MFCD00011340 |
InChI Key | NBIIXXVUZAFLBC-UHFFFAOYSA-N |
Synonym | orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico |
PubChem CID | 1004 |
ChEBI | CHEBI:26078 |
IUPAC Name | phosphoric acid |
SMILES | OP(=O)(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15667150
|
Honeywell-Fluka
79614-100ML |
100 mL |
£112.75
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15677150
|
Honeywell-Fluka
79614-500ML |
500 mL |
£405.76
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TraceSELECT™ Ultra and TraceSELECT
- ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions
Chemical Identifiers
7664-38-2 | |
97.994 | |
NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
1004 | |
phosphoric acid |
H3O4P | |
MFCD00011340 | |
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico | |
CHEBI:26078 | |
OP(=O)(O)O |
Specifications
7664-38-2 | |
1.685g/mL at 25°C | |
H3O4P | |
MFCD00011340 | |
1921286 | |
NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
phosphoric acid | |
1004 | |
98g/mol | |
Phosphoric acid |
85% | |
5 mmHg (25°C), 2.2 mmHg (20°C) | |
H3PO4 | |
UN1805 | |
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico | |
OP(=O)(O)O | |
97.994 | |
CHEBI:26078 | |
TraceSELECT™ |
Safety and Handling
P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
missing translation for 'rtecsNumber' : TB6300000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Phosphoric acid
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Orthophosphoric acid