missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric Acid - Triethylamine 1:1 Solution, Honeywell™ Fluka™
Click to view available options
Quantity:
100 mL
Packaging:
Glass Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS | 35365-94-7 |
Density | 1.09g/mL at 20°C |
Packaging | Glass Bottle |
Quantity | 100 mL |
Molecular Formula | C6H18NO4P |
Linear Formula | N(CH2(CH3)3 · H3PO4 |
Physical Form | Liquid |
MDL Number | MFCD00067475 |
UN Number | NONH for all modes of transport |
Synonym | triethylamine phosphate, triethylammonium phosphate, triethylamine phosphate 1:1, n,n-diethylethanamine; phosphoric acid, phosphoric acid; triethylamine, ethanamine, n,n-diethyl-, phosphate, ethanamine, n,n-diethyl-, phosphate 1:?, ethanamine, n,n-diethyl-, phosphate 1:1, teap, acmc-20aj7n |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'