Learn More
Phosphorus(III) bromide, 99%, Thermo Scientific Chemicals
Description
Phosphorus(III) bromide is used mainly for the conversion of alcohols to alkyl bromides and it is used in tests for skin corrosivity. It is used for alkene preparation and it act as a lewis base and a lewis acid. It is used as a catalyst for the alpha-bromination of carboxylic acids and used as intermediates in hell-volhard-zelinsky halogenation. Phosphorus tribromide is used in the manufacture of pharmaceuticals such as alprazolam, methohexital and fenoprofen. It is also used as a fire suppression agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 172.9°C |
Melting Point | -40°C |
Color | Colorless |
Physical Form | Liquid |
Quantity | 100 g |
Assay Percent Range | 99% (Metals Basis) |
UN Number | 1808 |
Beilstein | 3903060 |
Solubility Information | Micible in acetone,carbon disulfide,hydrogen sulfide,chloroform,diethyl ether and decomposes in water and alcohol. |
Formula Weight | 270.7 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
S20K042 | Certificate of Analysis | 03/07/2024 | 040191.14,040191.22 |
Y05H045 | Certificate of Analysis | 12/04/2022 | 040191.14,040191.22 |
Y05H048 | Certificate of Analysis | 12/04/2022 | 040191.14,040191.22 |
Y05H043 | Certificate of Analysis | 12/04/2022 | 040191.14,040191.22 |
Y05H042 | Certificate of Analysis | 12/04/2022 | 040191.14,040191.22 |
Safety and Handling


- Phosphorus(III) bromide
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- EUH014-Reacts violently with water.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Phosphorus tribromide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.