missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus(III) chloride, 98% min, Thermo Scientific Chemicals

Chemical Identifiers

CAS 7719-12-2
Molecular Formula Cl3P
Molecular Weight (g/mol) 137.32
MDL Number MFCD00011438
InChI Key FAIAAWCVCHQXDN-UHFFFAOYSA-N
Synonym phosphorus trichloride, phosphorous trichloride, trichlorophosphine, phosphorus chloride, phosphorous chloride, phosphorus iii chloride, pcl3, phosphorus chloride pcl3, phosphine, trichloro, fosfortrichloride
PubChem CID 24387
ChEBI CHEBI:30334
IUPAC Name trichlorophosphane
SMILES P(Cl)(Cl)Cl
View More Specs

Description

Description

Phosphorus(III) chloride is used as a reagent in organic synthesis. It is utilized in the conversion of primary alcohol and secondary alcohol to alkyl chlorides. It acts as a precursor to phosphorous pentachloride, phosphorous oxychloride, thiophosphoryl chloride and triphenylphosphine. It is also used to prepare phosphite esters, which is used Horner-Wadsworth-Emmons reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

Keep container tightly closed and dry. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H300+H330-H314-H373

missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: I

missing translation for 'einecsNumber' : 231-749-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phosphorus(III) chloride, 98% min, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.