Learn More
Phosphorus(III) chloride, 99.997% (metals basis), Thermo Scientific Chemicals
Chemical Identifiers
CAS | 7719-12-2 |
---|---|
Molecular Formula | Cl3P |
Molecular Weight (g/mol) | 137.32 |
MDL Number | MFCD00011438 |
InChI Key | FAIAAWCVCHQXDN-UHFFFAOYSA-N |
Synonym | phosphorus trichloride, phosphorous trichloride, trichlorophosphine, phosphorus chloride, phosphorous chloride, phosphorus iii chloride, pcl3, phosphorus chloride pcl3, phosphine, trichloro, fosfortrichloride |
PubChem CID | 24387 |
ChEBI | CHEBI:30334 |
IUPAC Name | trichlorophosphane |
SMILES | P(Cl)(Cl)Cl |
Description
Phosphorus(III) chloride is the precursor to organ phosphorus compounds, most notably phosphites and phosphonates. It is used during electrode position of metal on rubber and for making surfactants, gasoline additives, dyestuffs, textile finishing agents, medicinal products, and other chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7719-12-2 | |
137.32 | |
FAIAAWCVCHQXDN-UHFFFAOYSA-N | |
24387 | |
trichlorophosphane |
Cl3P | |
MFCD00011438 | |
phosphorus trichloride, phosphorous trichloride, trichlorophosphine, phosphorus chloride, phosphorous chloride, phosphorus iii chloride, pcl3, phosphorus chloride pcl3, phosphine, trichloro, fosfortrichloride | |
CHEBI:30334 | |
P(Cl)(Cl)Cl |
Specifications
76°C | |
7719-12-2 | |
Cl3P | |
UN1809 | |
phosphorus trichloride, phosphorous trichloride, trichlorophosphine, phosphorus chloride, phosphorous chloride, phosphorus iii chloride, pcl3, phosphorus chloride pcl3, phosphine, trichloro, fosfortrichloride | |
FAIAAWCVCHQXDN-UHFFFAOYSA-N | |
trichlorophosphane | |
24387 | |
137.33 | |
Acrid | |
Moisture sensitive | |
(metals basis) |
-111.8°C | |
Liquid | |
MFCD00011438 | |
14,7358 | |
Soluble in benzene,carbon sulfide,ether,chloroform,carbon tetrachloride. | |
P(Cl)(Cl)Cl | |
137.32 | |
CHEBI:30334 | |
99.997% | |
1.516 | |
1.574g/mL at 21°C | |
Phosphorus(III) chloride |
Safety and Handling
GHS H Statement
H300-H330-H314-H318-H373
Fatal if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H300+H330-H314-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: PHOSPHORUS TRICHLORIDE
missing translation for 'einecsNumber' : 231-749-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only