missing translation for 'onlineSavingsMsg'
Learn More
Phosphorus oxybromide, 95%, Thermo Scientific Chemicals Product Code.: 10035033

Phosphorus oxybromide, 95%, Thermo Scientific Chemicals

Product Code. 10035033
25 g, Glass bottle
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10035033

Brand: Thermo Scientific 388450250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-59-5
Molecular Formula Br3OP
Molecular Weight (g/mol) 286.69
InChI Key UXCDUFKZSUBXGM-UHFFFAOYSA-N
Synonym phosphorus oxybromide, phosphoric tribromide, phosphoryl bromide, phosphoryl tribromide, phosphorusoxybromide, phosphorus v oxybromide, unii-h98h8y87qq, phosphorousoxybromide, phosphoroyl tribromide, pobr3
PubChem CID 24613
SMILES O=P(Br)(Br)Br

Specifications

Boiling Point 192.0°C
Melting Point 56.0°C
Color Brown-Gray to Colorless
Physical Form Crystals or Fused Mass
Quantity 25 g
Assay Percent Range 94% min. (Argentometry)
Merck Index 15,746
Formula Weight 286.69
Percent Purity 95%
Packaging Glass bottle
Chemical Name or Material Phosphorus oxybromide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phosphorus oxybromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H290-May be corrosive to metals.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phosphorus oxybromide, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.