missing translation for 'onlineSavingsMsg'
Learn More
Phosphorus pentabromide, 95%, Thermo Scientific Chemicals Product Code.: 10020598

Phosphorus pentabromide, 95%, Thermo Scientific Chemicals

Product Code. 10020598
25 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10020598

Brand: Thermo Scientific Acros 428270250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-69-7
Molecular Formula Br5P
Molecular Weight (g/mol) 430.49
MDL Number MFCD00011437
InChI Key QRKVRHZNLKTPGF-UHFFFAOYSA-N
Synonym phosphorus pentabromide, phosphorane, pentabromo, unii-3d9wis0bqw, pentabromophosphorane, phosphorus v bromide, 3d9wis0bqw, pentabromo, phosphorpentabromid, phosphoruspentabromide, phosphorouspentabromideShow MoreShow Less
PubChem CID 62678
IUPAC Name pentabromo-$l^{5}-phosphane
SMILES P(Br)(Br)(Br)(Br)Br

Specifications

Melting Point 105°C to 107°C
Quantity 25 g
Assay Percent Range 94% min. (Argentometry)
Linear Formula PBr5
Merck Index 15,7462
Solubility Information Solubility in water: reacts
Formula Weight 430.49
Percent Purity 95%
Packaging Glass bottle
Chemical Name or Material Phosphorus pentabromide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462060 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number428271000,428270250
Lot NumberA0456464 Certificate TypeCertificate of Analysis Date03/05/2024 Catalog Number428271000,428270250
Lot NumberA0462060 Certificate TypeCertificate of Analysis Date19/04/2024 Catalog Number428271000,428270250
Lot NumberA0451459 Certificate TypeCertificate of Analysis Date09/10/2023 Catalog Number428271000,428270250
Lot NumberA0455427 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number428271000,428270250
compliance-icons
Product Identifier
  • Phosphorus pentabromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phosphorus pentabromide, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.