Learn More
Phosphorus pentasulfide, 98+%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.

Specifications
Specifications
Boiling Point | 514.0°C |
Melting Point | 282.0°C to 286.0°C |
Color | Yellow to Green |
Physical Form | Powder |
Quantity | 500 g |
Assay Percent Range | 98% min |
Linear Formula | P4S10 |
Fieser | 01,870; 03,226; 04,389; 05,534; 06,470; 07,290; 08,401; 09,374; 10,320 |
Flash Point | >100°C |
Merck Index | 15,7466 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0466993 | Certificate of Analysis | 23/10/2024 | 196725000,196720010 |
A0465926 | Certificate of Analysis | 04/09/2024 | 196725000,196720010 |
A0464376 | Certificate of Analysis | 08/07/2024 | 196725000,196720010 |
A0458448 | Certificate of Analysis | 12/12/2023 | 196725000,196720010 |
A0457250 | Certificate of Analysis | 24/10/2023 | 196725000,196720010 |
Safety and Handling



- Phosphorus pentasulfide
- Danger
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- Flammable solid Category 1
- Substance which\, in contact with water\, emits flammable gases Category 1
- H400-Very toxic to aquatic life.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H228-Flammable solid.
- EUH029-Contact with water liberates toxic gas.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.