missing translation for 'onlineSavingsMsg'
Learn More
Phosphorus pentasulfide, 98+%, Thermo Scientific Chemicals Product Code.: 10703011

Phosphorus pentasulfide, 98+%, Thermo Scientific Chemicals

Product Code. 10703011
1 kg, Glass bottle
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10703011

Brand: Thermo Scientific Acros 196720010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-80-3
Molecular Formula P4S10
Molecular Weight (g/mol) 444.48
MDL Number MFCD00011441
InChI Key CYQAYERJWZKYML-UHFFFAOYSA-N
Synonym phosphorus pentasulfide, sulfur phosphide, diphosphorus pentasulfide, diphosphorus pentasulphide, tetraphosphorus decasulfide, phosphoric sulfide, phosphorus persulfide, phosphorus pentasulphide, thiophosphoric anhydride, sirnik fosforecny czech
PubChem CID 14817
SMILES P12(=S)SP3(=S)SP(=S)(S1)SP(=S)(S2)S3

Specifications

Boiling Point 514.0°C
Melting Point 282.0°C to 286.0°C
Color Yellow to Green
Physical Form Powder
Quantity 1 kg
Assay Percent Range 98% min
Linear Formula P4S10
Fieser 01,870; 03,226; 04,389; 05,534; 06,470; 07,290; 08,401; 09,374; 10,320
Flash Point >100°C
Merck Index 15,7466
Solubility Information Solubility in water: reacts forming h3po4 and h2s. Other solubilities: soluble in solutions of alkali hydroxides, soluble in carbon disulfide, reacts with alcohols and acids
Formula Weight 444.48
Percent Purity ≥98%
Packaging Glass bottle
Chemical Name or Material Phosphorus pentasulfide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phosphorus pentasulfide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Flammable solid Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H228-Flammable solid.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phosphorus pentasulfide, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.