missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals

£23.81 - £358.51

Chemical Identifiers

CAS 1314-56-3
Molecular Formula O5P2
Molecular Weight (g/mol) 141.94
MDL Number MFCD00011440
InChI Key DLYUQMMRRRQYAE-UHFFFAOYSA-N
Synonym Phosphoric anhydride
IUPAC Name tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone
SMILES O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10340994
SDS View Documents
Thermo Scientific Acros
200892500
250 g Glass bottle
£23.81
250g
Please to purchase this item. Need a web account? Register with us today!
10124360
SDS View Documents
Thermo Scientific Acros
200890010
1 kg Glass bottle
£62.72
1kg
Please to purchase this item. Need a web account? Register with us today!
10217483
SDS View Documents
Thermo Scientific Acros
200890025
2.5 kg Glass bottle
£113.57
2.50kg
Please to purchase this item. Need a web account? Register with us today!
11994091
SDS View Documents
Thermo Scientific Acros
200890100
10 kg Plastic drum
£358.51
10kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.