missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus powder, -100 mesh, red amorphous, 98% (metals basis), Thermo Scientific Chemicals

Specifications

CAS 7723-14-0
Physical Form Solid
Percent Purity 98.9%
View More Specs

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

7723-14-0
Solid
98.9%
Videos
SDS
Documents

Documents

Product Certifications