missing translation for 'onlineSavingsMsg'
Learn More
Phosphorus solution for ICP 10g/L H2O matrix, Fisher chemical™ Product Code.: 12916364

Phosphorus solution for ICP 10g/L H2O matrix, Fisher chemical™

Product Code. 12916364
250 mL, Bottle
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Packaging:
Bottle
This item is not returnable. View return policy

Product Code. 12916364

Brand: Fisher Chemical S 240 250

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Bottle
Quantity 250 mL
Concentration 10±0.02 g/L
Type Mass Spectrometry Standard
Element P
Solvent or Matrix H2O

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1024101 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberS 240 250
Lot Number1023643 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberS 240 250
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phosphorus solution for ICP 10g/L H2O matrix, Fisher chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.