missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus Solution for ICP, 1g/L, H2O Matrix, Fisher Chemical™

Product Code. 12983464
Click to view available options
Quantity:
100 mL
250 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Concentration 1g/L
Quantity 250 mL
Solution Contains H2O
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'