missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphorus Standard in Base Oil 75, 1000μg/g, SPEX CertiPrep™
Click to view available options
Quantity:
50 g
Packaging:
Triple-leached LDPE bottle
Beschreibung
Beinhaltet
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
P | 1000μg/g |
Zertifizierungen
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Spezifikation
Spezifikation
CAS | 7723-14-0, 8042-47-5 |
CAS Min % | 0.5, 99.5 |
CAS Max % | 0.5, 99.5 |
Molecular Formula | P |
MDL Number | MFCD00133771 |
InChI Key | OAICVXFJPJFONN-UHFFFAOYSA-N |
SMILES | [P] |
IUPAC Name | phosphorus |
Molecular Weight (g/mol) | 30.97 |
PubChem CID | 5462309 |
Mehr anzeigen |
Product intended for laboratory use only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'