missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus tribromide, 98%

Product Code. 11303187
Click to view available options
Quantity:
25 g
100 g
1000 g
Unit Size:
100g
1kg
25g
This item is not returnable. View return policy

Product Code. 11303187

Brand: Thermo Scientific Alfa Aesar L13638.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-60-8 | Br{3}P

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 175°C
Melting Point -40°C
CAS 7789-60-8
Physical Form Liquid
Quantity 1000 g
Molecular Formula Br{3}P
Synonym Phosphorus(III) bromide
Percent Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.