missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus(V) bromide, 95%, Thermo Scientific Chemicals

Product Code. 11410667
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Phosphorus(V) bromide, is used in organic chemistry to convert carboxylic acids to acyl bromides

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phosphorus(V) bromide, is used in organic chemistry to convert carboxylic acids to acyl bromides

Solubility
Reacts with water.

Notes
Moisture Sensitive. Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 7789-69-7
Molecular Formula Br5P
Molecular Weight (g/mol) 430.494
MDL Number MFCD00011437
InChI Key QRKVRHZNLKTPGF-UHFFFAOYSA-N
Synonym phosphorus pentabromide, phosphorane, pentabromo, unii-3d9wis0bqw, pentabromophosphorane, phosphorus v bromide, 3d9wis0bqw, pentabromo, phosphorpentabromid, phosphoruspentabromide, phosphorouspentabromideShow MoreShow Less
PubChem CID 62678
IUPAC Name pentabromo-$l^{5}-phosphane
SMILES P(Br)(Br)(Br)(Br)Br

Specifications

Boiling Point 105°C to 107°C (decomposition)
Melting Point ∽105°C (decomposition)
Quantity 100 g
UN Number UN2691
Beilstein 3903332
Merck Index 14,7350
Solubility Information Reacts with water.
Formula Weight 430.5
Percent Purity 95%
Odor Acrid
Sensitivity Moisture sensitive
Chemical Name or Material Phosphorus(V) bromide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX25F002 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22481.14,B22481.36,B22481.22
Lot Number10146478 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22481.14,B22481.36,B22481.22
Lot NumberUSLY000651 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22481.14,B22481.36,B22481.22
Lot NumberR19H007 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22481.14,B22481.36,B22481.22
Lot NumberQ10E041 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22481.14,B22481.36,B22481.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phosphorus(V) bromide, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.