missing translation for 'onlineSavingsMsg'
Learn More
Phosphorus(V) oxide, 99.99%, Thermo Scientific Chemicals Product Code.: 11413880

Phosphorus(V) oxide, 99.99%, Thermo Scientific Chemicals

Product Code. 11413880
500 g
Click to view available options
Quantity:
10 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11413880

Brand: Thermo Scientific Alfa Aesar 089966.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Phosphorus(V) oxide is used as a drying and dehydrating agent, a condensation reagent in organic synthesis and a laboratory reagent. It is also used in sugar refining and in fire extinguishing. Phosphorus pentoxide in DMSO forms an Onodera reagent which oxidizes alcohols. It is capable of converting mineral acids to anhydrides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-56-3
Molecular Formula O5P2
Molecular Weight (g/mol) 141.94
MDL Number MFCD00011440
InChI Key DLYUQMMRRRQYAE-UHFFFAOYSA-N
IUPAC Name tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone
SMILES O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3

Specifications

Boiling Point 360°C (sublimation)
Melting Point 340°C
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.99%
Solubility Information Soluble in sulfuric acid. Insoluble in acetone and ammonia. Decomposes in water.
Formula Weight 141.94
Odor Odorless
Sensitivity Moisture sensitive
Density 2.39
Chemical Name or Material Phosphorus(V) oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY14D035 Certificate TypeCertificate of Analysis Date13/01/2025 Catalog Number089966.22,089966.36,089966.09
Lot NumberR07E005 Certificate TypeCertificate of Analysis Date13/01/2025 Catalog Number089966.22,089966.36,089966.09
Lot NumberQ28G036 Certificate TypeCertificate of Analysis Date13/01/2025 Catalog Number089966.22,089966.36,089966.09
Lot NumberT29I012 Certificate TypeCertificate of Analysis Date20/09/2022 Catalog Number089966.22,089966.36,089966.09
Lot NumberF11Z013 Certificate TypeCertificate of Analysis Date02/08/2022 Catalog Number089966.22,089966.36,089966.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phosphorus(V) oxide, 99.99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.