missing translation for 'onlineSavingsMsg'
Learn More
Phosphorus(V) oxide, ACS, 98% min, Thermo Scientific Chemicals Product Code.: 11386038

Phosphorus(V) oxide, ACS, 98% min, Thermo Scientific Chemicals

Product Code. 11386038
500 g
Click to view available options
Quantity:
50 g
500 g
4 x 500 g
This item is not returnable. View return policy

Product Code. 11386038

Brand: Thermo Scientific Alfa Aesar 010524.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Use As a Drying and Dehydrating Agent, As a Condensing Agent in organic Synthesis, in Sugar Refining

Used as a drying and dehydrating agent, condensation reagent in organic synthesis and a laboratory reagent. Used in sugar refining, and in fire extinguishing. Phosphorus pentoxide in DMSO forms Onodera reagent, which oxidizes alcohols. Capable of converting mineral acids to anhydrides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-56-3
Molecular Formula O5P2
Molecular Weight (g/mol) 141.94
MDL Number MFCD00011440
InChI Key DLYUQMMRRRQYAE-UHFFFAOYSA-N
IUPAC Name tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone
SMILES O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3

Specifications

Boiling Point 360°C (sublimation)
Melting Point 340°C
Color White
Physical Form Powder
Quantity 500 g
UN Number UN1807
Merck Index 14,7355
Synonym Phosphoric anhydride; Phosphoric oxide
Solubility Information Soluble in H2SO4; Insoluble in acetone and ammonia; Decomposes in water.
Formula Weight 141.94
Percent Purity ≥98%
Grade ACS
Odor Weak
Sensitivity Moisture sensitive
Density 2.39
Chemical Name or Material Phosphorus(V) oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phosphorus(V) oxide, ACS, 98% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.