missing translation for 'onlineSavingsMsg'
Learn More

Phosphotungstic acid hydrate, Thermo Scientific Chemicals

Product Code. 10781861
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12501-23-4
Molecular Formula H5O41PW12
Molecular Weight (g/mol) 2880.20
MDL Number MFCD00011342
InChI Key AVFBYUADVDVJQL-UHFFFAOYSA-N
Synonym phosphotungstic acid hydrate, tungstophosphoric acid, 12-tungstophosphoric acid hydrate, phosphoric acid dodecakis tungsten trioxide hydrate, iuaxea, aronis24073, phosphotungstic acid hydrate, reagent grade, phosphotungstic acid hydrate, for microscopy, phosphotungstic acid hydrate, p.a, phosphotungstic acid hydrate, saj special grade
PubChem CID 16212977
SMILES O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.OP(O)(O)=O

Specifications

Packaging Plastic bottle
Quantity 500 g
Linear Formula 12WO3·xH2O
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone, diethyl ether and alcohol
IUPAC Name phosphoric acid;trioxotungsten;hydrate
Formula Weight 2880.17
Chemical Name or Material Phosphotungstic acid hydrate, p.a.
compliance-icons
Product Identifier
  • Phosphotungstic acid hydrate
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'