missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphotungstic Acid, n-Hydrate, Crystal, BAKER ANALYZED™ Reagent, J.T. Baker™
Click to view available options
Quantity:
125 g
Packaging:
Poly Bottle
Specifications
Specifications
CAS | 12067-99-1 |
Packaging | Poly Bottle |
Molecular Formula | P2O5·24WO3·nH2O |
Quantity | 125 g |
Specific Gravity | 1L = 2.13kg |
Grade | BAKER ANALYZED™ Reagent |
Physical Form | Crystals |
Chemical Name or Material | Phosphotungstic Acid, n-Hydrate |
Safety and Handling
Signal Word
- Danger
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'