Learn More
Lovibond™ Tintometer™ Photometer-System PC MultiDirect
Used in Waste Water, Drinking Water, Industrial Process Water, Scientific Applications, Governmental and Private Laboratories, Universities and Research Centres, Mobile Applications
Brand:  Lovibond™ 210000
Product Code. 10089841
  • £1380.00 / Each
Explore more special offers
Description
Description
  • Dual Beam Technology and Interference Filters for highest accuracy
  • A wide range of pre-programmed methods
  • Long-term stable LEDs as light sources
  • Update of new methods and languages via Internet (free of charge)
  • Interface
  • Memory for 1000 data sets
  • Mobile
Specifications
Specifications
Photometer-System PC Multidirect
LED
Waste Water, Drinking Water, Industrial Process Water, Scientific Applications, Governmental and Private Laboratories, Universities and Research Centres, Mobile Applications
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lovibond™ Tintometer™ Photometer-System PC MultiDirect >