missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHOX2B Mouse anti-Human, Mouse, Clone: 1F5F5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66254-1-IG-150UL
This item is not returnable.
View return policy
Description
The DNA-associated protein encoded by this gene is a member of the paired family of homeobox proteins localized to the nucleus. The protein functions as a transcription factor involved in the development of several major noradrenergic neuron populations and the determination of neurotransmitter phenotype. The gene product is linked to enhancement of second messenger-mediated activation of the dopamine beta-hydroylase, c-fos promoters and several enhancers, including cyclic amp-response element and serum-response element.Specifications
PHOX2B | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35690, Q99453 | |
Phox2b | |
PHOX2B Fusion Protein Ag13196 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
1F5F5 | |
Unconjugated | |
PHOX2B | |
NBPhox, Neuroblastoma Phox, paired like homeobox 2b, PHOX2B, PHOX2B homeodomain protein, PMX2B | |
Mouse | |
Protein A | |
RUO | |
18935, 8929 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction