missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ pHrodo™ Red, succinimidyl ester (pHrodo™ Red, SE)

Product Code. 10676983
Click to view available options
Quantity:
1 mg
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10676983

Brand: Molecular Probes™ P36600

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

New pH-sensitive pHrodo™ Red dye conjugates give faster and more accurate results than any other phagocytosis assay.

New pH-sensitive pHrodo™ Red dye conjugates give faster and more accurate results than any other phagocytosis assay

pHrodo™ Red dye conjugates are non-fluorescent outside the cell, but fluoresce brightly red in phagosomes

Get faster staining and more accurate results - without the need for wash steps or quencher dye

• Specific detection of phagocytosis and endocytosis
• Reduced signal variability and improved timing in sensitive experiments
• Multiplex with green dyes such as GFP, Fluo-4, or calcein

The fluorescence of the novel pHrodo™ Red dye dramatically increases as pH decreases from neutral to the acidic, making it an ideal tool to study phagocytosis and its regulation by drugs and/or environmental factors. The lack of fluorescence outside the cell eliminates the need for wash steps and quencher dyes.

Use the ready-made pHrodo™ Red E.coli BioParticles™ conjugates in imaging or flow applications, or pHrodo™ Red SE, the activated succinimidyl ester, for labeling microorganisms or proteins of your choice.

Consult user Manual for solubility instructions.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Product Type Succinimidyl Ester
Molecular Weight (g/mol) ∼650
Content And Storage Store in freezer (-5°C to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye pHrodo™ Red
Label Type Classic Dyes
Color Red
Product Line pHrodo
Emission 587 nm
Excitation 560 nm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.