missing translation for 'onlineSavingsMsg'
Learn More
Phthalazine, 98%, Thermo Scientific Chemicals Product Code.: 11464983

Phthalazine, 98%, Thermo Scientific Chemicals

Product Code. 11464983
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11464983

Brand: Thermo Scientific Alfa Aesar A12270.14

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Phthalazine derivatives including copper and platinum complexes were induce the apoptosis in breast cancer cell lines and renal hyperemia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthalazine derivatives including copper and platinum complexes were induce the apoptosis in breast cancer cell lines and renal hyperemia.

Solubility
Soluble in water, methanol, chloroform.

Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 253-52-1
Molecular Formula C8H6N2
Molecular Weight (g/mol) 130.15
MDL Number MFCD00006908
InChI Key LFSXCDWNBUNEEM-UHFFFAOYSA-N
Synonym 2,3-benzodiazine, benzo d pyridazine, 2,3-diazanaphthalene, beta-phenodiazine, .beta.-phenodiazine, unii-91y28dm24n, ccris 6894, phthalazin, benzo-orthodiazine
PubChem CID 9207
ChEBI CHEBI:36597
IUPAC Name phthalazine
SMILES C1=CC=C2C=NN=CC2=C1

Specifications

Melting Point 89°C to 92°C
Boiling Point 317°C
Quantity 25 g
Beilstein 109240
Merck Index 14,7370
Solubility Information Soluble in water,methanol,chloroform.
Formula Weight 130.15
Percent Purity 98%
Chemical Name or Material Phthalazine
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phthalazine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.