missing translation for 'onlineSavingsMsg'
Learn More
Phthalic acid, ACS, 99.5+%, Thermo Scientific Chemicals Product Code.: 11348918

Phthalic acid, ACS, 99.5+%, Thermo Scientific Chemicals

Product Code. 11348918
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11348918

Brand: Thermo Scientific Alfa Aesar 041770.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Phthalic acid is used as an active ingredient in polyesters and alkyd resins. It is also used as plasticizers for polyvinyl chloride and other polymers. It is also useful in the production of anthraquinone, phenolphthalein and phthalocyanine pigments. It serves as an organic reagent in the production of phthalates. It is used to prepare 1,3-cyclohexadiene by reduction using sodium amalgam.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-99-3
Molecular Formula C8H6O4
Molecular Weight (g/mol) 166.132
MDL Number MFCD00002467
InChI Key XNGIFLGASWRNHJ-UHFFFAOYSA-N
Synonym 1,2-benzenedicarboxylic acid, benzene-1,2-dicarboxylic acid, o-phthalic acid, o-dicarboxybenzene, o-benzenedicarboxylic acid, pathalic acid, acide phtalique, kyselina ftalova, ortho-phthalic acid, phthalicacidShow MoreShow Less
PubChem CID 1017
ChEBI CHEBI:29069
IUPAC Name phthalic acid
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)O

Specifications

Melting Point 205°C (decomposition)
Assay Percent Range >99.5%
Beilstein 608199
Merck Index 14,7371
Solubility Information Soluble in alcohol. Slightly soluble in water,methanol and ether. Insoluble in chloroform.
Formula Weight 166.13
Percent Purity ≥99.5%
Grade ACS
Density 1.593
Quantity 100 g
Flash Point 168°C (334°F)
Physical Form Crystalline Powder
Chemical Name or Material Phthalic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN24L031 Certificate TypeCertificate of Analysis Date26/02/2025 Catalog Number041770.22,041770.36
Lot NumberW10K040 Certificate TypeCertificate of Analysis Date23/09/2024 Catalog Number041770.22,041770.36
Lot NumberS14K021 Certificate TypeCertificate of Analysis Date28/06/2024 Catalog Number041770.22,041770.36
Lot NumberQ23K046 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number041770.22,041770.36
Lot NumberP06K043 Certificate TypeCertificate of Analysis Date15/03/2024 Catalog Number041770.22,041770.36
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phthalic acid, ACS, 99.5+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.